ADV371426459 AChemBlock

Methyl 3-iodo-1H-indazole-4-carboxylate

  • MDL number MFCD07781588
  • Cas Number : 885521-54-0
  • Molecular Weight : 302.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-iodo-1h-indazole-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2043
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart