ADV371426505 AChemBlock

2-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

  • MDL number MFCD11109457
  • Cas Number : 845751-67-9
  • Molecular Weight : 258.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methylindazole-7-boronic-acid-pinacol-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2089
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart