ADV371426516 AChemBlock

7-Bromo-3-iodoindazole

  • MDL number MFCD08689979
  • Cas Number : 944904-26-1
  • Molecular Weight : 322.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-3-iodoindazole.html

Safety Information

signalword warning
pcodes P261;P271;P280
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2100
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart