ADV371426519 AChemBlock

5-Iodoindazole

  • MDL number MFCD07781642
  • Cas Number : 55919-82-9
  • Molecular Weight : 244.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form beige powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-iodoindazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2103
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart