ADV371426525 AChemBlock

2,3-Dimethyl-6-nitro-1H-indazole

  • MDL number MFCD10703326
  • Cas Number : 444731-73-1
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-3-dimethyl-6-nitro-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2109
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart