ADV371426529 AChemBlock

2-Methylindazole-6-carboxylic acid

  • MDL number MFCD15071449
  • Cas Number : 1031417-46-5
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methylindazole-6-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2113
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart