ADV371426557 AChemBlock

Methyl 5-nitro-1H-indazole-3-carboxylate

  • MDL number MFCD07371594
  • Cas Number : 78155-75-6
  • Molecular Weight : 221.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-nitro-1h-indazole-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2145
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart