ADV371426560 AChemBlock

3-Methylenecyclobutanecarboxylic acid

  • MDL number MFCD00101901
  • Cas Number : 15760-36-8
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-methylenecyclobutanecarboxylic-acid.html

Safety Information

signalword Danger
pcodes P261;P264;P271;P280;P302+352;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2502
pictograms GHS05; GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart