ADV371426568 AChemBlock

2-Methylpiperidin-4-one hydrochloride

  • MDL number MFCD08669336
  • Cas Number : 13729-77-6
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methylpiperidin-4-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2514
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart