ADV371426579 AChemBlock

5-Bromoisoindolin-1-one

  • MDL number MFCD09701292
  • Cas Number : 552330-86-6
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromoisoindolin-1-one.html

Safety Information

signalword Danger
pcodes P264;P270;P301+310;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2558
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart