ADV371426586 AChemBlock

3-Hydroxymethyl cyclobutane-one

  • MDL number MFCD11100969
  • Cas Number : 183616-18-4
  • Molecular Weight : 100.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light-yellow liquid or white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxymethyl-cyclobutane-one.html

Safety Information

signalword Warning
pcodes P261;P264;P270;P271;P280;P301+312;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2565
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart