ADV371426595 AChemBlock

tert-Butyl 4-carbamothioylpiperazine-1-carboxylate

  • MDL number MFCD07787249
  • Cas Number : 196811-66-2
  • Molecular Weight : 245.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-carbamothioylpiperazine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2704
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart