ADV371426599 AChemBlock

1,4-Diazabicyclo[3.2.2]nonan-3-one

  • MDL number MFCD01815127
  • Cas Number : 53619-11-7
  • Molecular Weight : 140.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-4-diazabicyclo-3-2-2-nonan-3-one.html

Safety Information

signalword Warning
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P337+313;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2716
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart