ADV371426602 AChemBlock

1-Boc-2,2-dimethylpiperazine

  • MDL number MFCD07371652
  • Cas Number : 674792-07-5
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-boc-2-2-dimethylpiperazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=D-2720
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart