ADV371426612 AChemBlock

4-Bromo-2-cyanopyridine

  • MDL number MFCD04065805
  • Cas Number : 62150-45-2
  • Molecular Weight : 183.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-cyanopyridine.html

Safety Information

signalword Warning
pcodes P262;P280;P302+352;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=E-3011
pictograms GHS06
hstatements H302;H319
Availability details for
Quantity
  • Availability
  • Add to Cart