ADV371426617 AChemBlock

4-Bromo-pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD07374897
  • Cas Number : 29681-42-3
  • Molecular Weight : 216.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-pyridine-2-carboxylic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=E-3030
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart