ADV371426631 AChemBlock

3-Cyano-5-hydroxypyridine

  • MDL number MFCD11044307
  • Cas Number : 152803-24-2
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form brown powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-cyano-5-hydroxypyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=E-3095
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart