ADV371426661 AChemBlock

6-(Bromomethyl)-2-methylquinoline

  • MDL number MFCD06658974
  • Cas Number : 141848-60-4
  • Molecular Weight : 236.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid/off-white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromomethyl-2-methylquinoline.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4030
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart