ADV371426693 AChemBlock

3-Cyano-4-iodopyridine

  • MDL number MFCD09037453
  • Cas Number : 490039-72-0
  • Molecular Weight : 230.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96.00
Storage temp. Store at Room Tem.
Physical Form yellow solid
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/3-cyano-4-iodopyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4078
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart