ADV371426696 AChemBlock

Toluene-4-sulfonic acid oxetan-2-ylmethyl ester

  • MDL number MFCD08544404
  • Cas Number : 115845-51-7
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/toluene-4-sulfonic-acid-oxetan-2-ylmethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4080
pictograms GHS07
hstatements H303;H335
Availability details for
Quantity
  • Availability
  • Add to Cart