ADV371426699 AChemBlock

cis-(3S,4R)-tert-Butyl 3-Amino-4-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD14582421
  • Cas Number : 138026-97-8
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3s-4r-tert-butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4084
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart