ADV371426706 AChemBlock

4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylic acid benzyl ester

  • MDL number MFCD11521562
  • Cas Number : 286961-15-7
  • Molecular Weight : 343.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-3-6-dihydro-2h-pyridine-1-carboxylic-acid-benzyl-ester.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4091
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart