ADV371426714 AChemBlock

(R)-2-Methylmorpholine

  • MDL number MFCD11041200
  • Cas Number : 790184-33-7
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form colorless or light yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-2-methylmorpholine.html
Availability details for
Quantity
  • Availability
  • Add to Cart