ADV371426715 AChemBlock

(R)-3-Ethylmorpholine

  • MDL number MFCD11519096
  • Cas Number : 74572-05-7
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form colorless or light yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-3-ethylmorpholine.html
Availability details for
Quantity
  • Availability
  • Add to Cart