ADV371426719 AChemBlock

(S)-3-Isopropylmorpholine

  • MDL number MFCD11519112
  • Cas Number : 77897-21-3
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at 0-8 �C
Physical Form colorless or light yellow liquid
Shipping Temperature Store at 0-8 �C
Country of Origin CN
Product Link https://www.achemblock.com/s-3-isopropylmorpholine.html
Availability details for
Quantity
  • Availability
  • Add to Cart