ADV371426721 AChemBlock

(S)-2-Ethylmorpholine

  • MDL number MFCD11519095
  • Cas Number : 74572-14-8
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form colorless or light yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-2-ethylmorpholine.html
Availability details for
Quantity
  • Availability
  • Add to Cart