ADV371426722 AChemBlock

(R)-2-Ethylmorpholine

  • MDL number MFCD14635494
  • Cas Number : 1313176-45-2
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at 0-8 �C
Physical Form colorless or light yellow liquid
Shipping Temperature Store at 0-8 �C
Country of Origin CN
Product Link https://www.achemblock.com/r-2-ethylmorpholine.html
Availability details for
Quantity
  • Availability
  • Add to Cart