ADV371426725 AChemBlock

(R)-2-Isopropylmorpholine

  • MDL number MFCD14582703
  • Cas Number : 792886-64-7
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form colorless or light yellow liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/r-2-isopropylmorpholine.html
Availability details for
Quantity
  • Availability
  • Add to Cart