ADV371426729 AChemBlock

2-Hydroxymethyl-morpholine hydrochloride

  • MDL number MFCD13191721
  • Cas Number : 144053-98-5
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-hydroxymethyl-morpholine-hydrochloride.html

Safety Information

signalword warning
pcodes P280;P261;P305+351+338;P302+350;P301+330+331
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1078
pictograms GHS05; GHS07
hstatements H302;H311;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart