ADV371426741 AChemBlock

3-Benzyl-morpholine

  • MDL number MFCD09991891
  • Cas Number : 7684-27-7
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form colorless or light yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-benzyl-morpholine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1090
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart