ADV371426742 AChemBlock

(S)-3-Benzyl-morpholine

  • MDL number MFCD11519115
  • Cas Number : 77897-23-5
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form colorless or light yellow liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/s-3-benzyl-morpholine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1091
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart