ADV371426744 AChemBlock

3-Phenyl-morpholine

  • MDL number MFCD09608120
  • Cas Number : 138713-44-7
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form colorless or light yellow liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-phenyl-morpholine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1093
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart