ADV371426766 AChemBlock

trans-3-Amino-1-Boc-4-hydroxypyrrolidine

  • MDL number MFCD06858457
  • Cas Number : 148214-90-8
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-amino-1-boc-4-hydroxypyrrolidine.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4114
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart