ADV371426769 AChemBlock

6-Amino-2-aza-spiro[3.4]octane-2-carboxylic acid tert-butyl ester

  • MDL number MFCD21648497
  • Cas Number : 1638763-38-8
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-amino-2-aza-spiro-3-4-octane-2-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4117
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart