ADV371426774 AChemBlock

2-Chloro-3-[1,3]dithian-2-yl-pyrazine

  • MDL number MFCD20926169
  • Cas Number : 874114-28-0
  • Molecular Weight : 232.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-3-1-3-dithian-2-yl-pyrazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4123
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart