ADV371426775 AChemBlock

6-Amino-2-aza-spiro[3.3]heptane-2,6-dicarboxylic acid 2-tert-butyl ester

  • MDL number MFCD20926180
  • Cas Number : 1363380-56-6
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-amino-2-aza-spiro-3-3-heptane-2-6-dicarboxylic-acid-2-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4124
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart