ADV371426784 AChemBlock

8-Bromo-1,2,3,4-tetrahydro-[1,6]naphthyridine

  • MDL number MFCD11847918
  • Cas Number : 362606-16-4
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-1-2-3-4-tetrahydro-1-6-naphthyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4134
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart