ADV371426788 AChemBlock

3-Hydroxymethyl-cyclobutanecarboxylic acid benzyl ester

  • MDL number MFCD13659349
  • Cas Number : 939768-59-9
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxymethyl-cyclobutanecarboxylic-acid-benzyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4138
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart