ADV371426791 AChemBlock

7-Amino-2-aza-spiro[3.5]nonane-2-carboxylic acid tert-butyl ester

  • MDL number MFCD21648502
  • Cas Number : 1408075-19-3
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-amino-2-aza-spiro-3-5-nonane-2-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4141
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart