ADV371426792 AChemBlock

4-Bromo-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD11040189
  • Cas Number : 30129-51-2
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4142
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart