ADV390216486 AChemBlock

2-Aza-tricyclo[3.3.1.13,7]decane-5-carboxylic acid methyl ester hydrochloride

  • MDL number MFCD19707044
  • Cas Number : 1389264-13-4
  • Molecular Weight : 231.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-aza-tricyclo-3-3-1-13-7-decane-5-carboxylic-acid-methyl-ester-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4082
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart