ADV390216489 AChemBlock

2-Aza-spiro[3.3]heptan-6-one hydrochloride

  • MDL number MFCD23703538
  • Cas Number : 1820703-11-4
  • Molecular Weight : 147.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-aza-spiro-3-3-heptan-6-one-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4151
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart