ADV390216494 AChemBlock

2-Acetylamino-4-bromo-benzamide

  • MDL number MFCD23703543
  • Cas Number : 1820704-22-0
  • Molecular Weight : 257.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-acetylamino-4-bromo-benzamide.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4222
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart