ADV390216496 AChemBlock

4-[2-Fluoro-1-(fluoromethyl)ethoxy]aniline hydrochloride

  • MDL number MFCD23703548
  • Cas Number : 109230-64-0
  • Molecular Weight : 223.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-2-fluoro-1-fluoromethyl-ethoxy-aniline-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4271
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart