ADV390216503 AChemBlock

3,3-Dimethyl-1-oxo-2,3-dihydro-1H-isoindole-5-carbonitrile

  • MDL number MFCD23703554
  • Cas Number : 1440519-98-1
  • Molecular Weight : 186.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethyl-1-oxo-2-3-dihydro-1h-isoindole-5-carbonitrile.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5043
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart