ADV390216504 AChemBlock

4-[[(1-Methylethyl)sulfonyl]methyl]-piperidine hydrochloride

  • MDL number MFCD23703555
  • Cas Number : 1018533-19-1
  • Molecular Weight : 241.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-1-methylethyl-sulfonyl-methyl-piperidine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4309
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart