ADV390216527 AChemBlock

N-(Tetrahydro-4-phenyl-2H-pyran-4-yl)acetamide

  • MDL number MFCD21641995
  • Cas Number : 939801-34-0
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-tetrahydro-4-phenyl-2h-pyran-4-yl-acetamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5761
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart