ADV390216530 AChemBlock

4-(3-Chloro-4-fiuoro-phenylamino)-7-fluoro-6-nitroquninazoline

  • MDL number MFCD09998825
  • Cas Number : 162012-67-1
  • Molecular Weight : 336.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-3-chloro-4-fiuoro-phenylamino-7-fluoro-6-nitroquninazoline.html

Safety Information

signalword Danger
pcodes P261;P264;P271;P280;P302+352;P304+340;P305+351+338;P310;P403+233
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3465
pictograms GHS05; GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart