ADV390216534 AChemBlock

2-Bromo-5-iodo-pyrazine

  • MDL number MFCD08061602
  • Cas Number : 622392-04-5
  • Molecular Weight : 284.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-5-iodo-pyrazine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3475
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart