ADV390216535 AChemBlock

4,5-Dichloro-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD16987934
  • Cas Number : 99469-85-9
  • Molecular Weight : 195.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-5-dichloro-2-methylsulfanyl-pyrimidine.html

Safety Information

signalword Danger
pcodes P210;P233;P240;P241;P242;P243;P264;P270;P280;P301+312;P303+361+353;P305+351+338;P310;P370+378;P403+235
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3509
pictograms GHS02; GHS05; GHS07
hstatements H226;H302;H318
Availability details for
Quantity
  • Availability
  • Add to Cart